3B Scientific® 6 Vertebrae Set
Supplier: American 3B Scientific
1000147
470118-952EA 149 USD
470118-952
3B Scientific® 6 Vertebrae Set
Models Human Biology Models
Represent Spinal Structure With A Compact Display
  • Vertebrae From Each Region Of The Spine
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
Consisting of atlas, axis, another cervical vertebra, two thoracic vertebrae with inter-vertebral discs and one lumbar vertebra. On stand, removable.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.