Solar LED Jar Light Kit (Soldering)
Supplier: KIDWIND PROJECT INC SE
900-10017-30-BL
470118-648CS 54.25 USD
470118-648
Solar LED Jar Light Kit (Soldering)
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Developed in collaboration with a science teacher from Pennsylvania, our build your own solar LED Jar light is a fun and creative way to teach and learn about solar electricity. Use your own mason jar or recycled jar and build a colorful LED light that automatically comes on at night and goes off at dawn. Comes with everything you need. This kit requires soldering. Tools needed are soldering iron, solder & flux, hot glue gun, electric drill, 3V 70mA Polycrystalline Solar Panel, Dusk/dawn circuit board, 2 Bright 5mm LED's BLUE, 2 AAA NiMH rechargeable batteries & Battery holder andMore Product Information