Science Take-Out® Taste Blind
Supplier: SCIENCE TAKE-OUT, LLC
STO-141U STO-141R STO-141 STO-141-NOTG
470118-360PK 113.5 USD
470118-360 470118-378 470118-380 470118-381
Science Take-Out® Taste Blind
Educational Materials Biology Educational Materials Animal Biology Learning Activites
Students use taste papers to determine if they are PTC tasters.
  • Available as a Single Kit with Material for One Student or Group
  • Unassembled 10 Pack for Additional Savings
  • No Additional Equipment Needed
  • Activities Include Interpreting a Pedigree and a Simulated Electrophoresis Gel
  • Students Determine the Genotypes of 10 Family Members
They also learn about the parts of the nervous system involved in tasting.
To gain this understanding, they interpret a pedigree and a simulated electrophoresis gel to determine the genotypes of 10 family members.
Ordering information: This complete experiment setup contains all materials and does not require lab equipment.