Polarizing Film, 6 × 6”
Supplier: United Scientific Supplies
PLF060
470117-788EA 50.5 USD
470117-788 470117-789
Polarizing Film, 6 × 6”
Educational Materials Physics Educational Materials Optics Learning Activities
Demonstrate how polarizing filters affect light.
  • Comes with a support stand
  • Comes as pairs to filter out light
Unmounted 6” x 6” film is an effective yet economical tool for demonstrating polarization of light. When the two filters are placed 90 degrees to each other, they block out a majority of the incoming light.