Vernier® Radiation Monitor
Supplier: Vernier Software & Technology
VRM-BTD
470116-638EA 198.99 USD
470116-638
Vernier® Radiation Monitor
Radiation Meters
Detect radiation and measure decay rates.
  • Requires no battery
  • X-ray, alpha, beta and gamma radiation can be detected
  • Red LED blinks on radiation detection
The Vernier Radiation Monitor consists of a Geiger-Mueller tube mounted in a small, rugged, plastic case. A thin window protected by a metal screen allows alpha radiation to be detected, along with beta and gamma. It can be used to explore radiation statistics, measure the rate of nuclear decay, and monitor radon progeny.
For educational use only.