Vernier® Resistor Board
Supplier: Vernier Software & Technology
VES-RB
470116-634EA 18.99 USD
470116-634
Vernier® Resistor Board
Power Supplies
The Vernier Resistor Board has seven 1 W 5% resistors mounted on a circuit board.
Test clip holes on either side of the resistors allow for easy connections to clips on wire leads.