Vernier® Melt Station
Supplier: Vernier Software & Technology
MLT-BTA MLT-TUBE
470113-968EA 628.99 USD
470113-968 470219-304
Vernier® Melt Station
Educational Materials Chemistry Educational Materials Applied Chemistry Learning Activities
Accurately determine the melting temperature of a solid.
  • Accurate results using built-in RTD over a range of 30°C to 260°C
  • Accuracy ± 0.31 + 0.0006T, where T is the temperature in Celsius
  • Resolution 0.10°C
  • High-quality 6X viewing lens for clear observation of the samples
  • Innovative, adjustable tilt allows for optimal viewing angle
  • Cooling fan reduces time between tests
Red LED (indicates heating mode)
Blue LED (indicates cooling mode with cooling fan running)
Yellow LED (safety shut off activated)
Connect the Melt Station to a Vernier interface to measure the melting temperature of solid substances.
For educational use only.
Ordering information: Each Melt Station comes with 100 standard capillary tubes. Additional packages of 100 are available.