Rotary Motion Motor Kit
Supplier: Vernier Software & Technology
MK-RMV
470113-966EA 11.99 USD
470113-966
Rotary Motion Motor Kit
Tachometers
This kit includes a small electric motor with pulley, rubber band belt, motor clip, and mounting screw. The motor can be attached to the Rotary Motion Sensor, and be used as an analog tachometer, or generator. You can also perform experiments that investigate motor efficiency in different conditions.