Microcentrifuge Tube Lid Locks
Supplier: United Scientific Supplies
145063
470112-976PK 1000 USD
470112-976
Microcentrifuge Tube Lid Locks
Closures Test/Sample Tube Closures
Secure caps to lids of microcentrifuge tubes with these autoclavable locks.
  • Easy to use tabs
  • Autoclavable
Each features a tab, allows to pick them out of a waterbath without burning your fingers.