30 V Voltage Probe
Supplier: Vernier Software & Technology
30V-BTA
470112-966EA 58.99 USD
470112-966
30 V Voltage Probe
Electrical Meters Voltmeters
This sensor is used to measure voltages in the range of -30 to +30 volts.
  • Voltage probe covers such a wide voltage range
  • Resolution is lower than our Differential Voltage Probe
Mainly used for experiments that involve voltages larger than 10 volts, such as when working with large solar panels.