Inductor
Supplier: Vernier Software & Technology
IND
470112-964EA 39.99 USD
470112-964
Inductor
Wire
This air-core inductor is made from 20 gauge copper wire.
It has an inductance of 5 mH and low resistance. A metal core can be added to increase inductance.