Green Diffraction Laser
Supplier: Vernier Software & Technology
GDL-DAK
470112-944EA 262.49 USD
470112-944
Green Diffraction Laser
Laser Pointers
The Green Diffraction Laser is an optional accessory for the Diffraction Apparatus.
Operating at 532 nm, it allows students to see and measure how diffraction patterns depend on light wavelength.