Vernier EasyTemp® Sensor
Supplier: Vernier Software & Technology
EZ-TMP
470112-924EA 50.39 USD
470112-924
Vernier EasyTemp® Sensor
Temperature Probes
Rugged, general-purpose temperature sensor for TI-84 Plus and TI-Nspire calculators.
  • Resolution 0.07 °C
  • Response time 4 s
Vernier EasyTemp® has a mini-A USB connector that allows you to attach the temperature probe directly to the USB port of a TI-84 Plus graphing calculator or TI-Nspire® handheld. Connecting the sensor to the calculator or handheld will automatically launch the data collection application, and you are ready to collect data.