Kick-Net
Supplier: LaMotte
0021-P
470112-698EA 143 USD
470112-698
Kick-Net
Samplers Water Samplers
Easily Collect Benthic Macroinvertebrates
  • GLOBE Approved
  • Easily Transportable
Use this Kick-Net to perform USEPA Rapid Bioassessment Protocols on benthic macroinvertebrates. Net is one meter square of 500 micron material. Contains hemmed sides and reinforced bottom seams to ensure anchoring. Complete with poles.