Riparian Component
Supplier: JT & A, INC.
71005RIP
470112-612EA 153.9 USD
470112-612
Riparian Component
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Expand Your Enviroscape® Model with the Following Components
Expand Your Enviroscape® Model with the Following Components:Expansion kit addresses the benefits and values of riparian buffers—areas including trees and other vegetation adjacent to the banks of streams, rivers and lakes. The User’s guide details five activities to demonstrate: nonpoint source pollution on sparse landscape, moderate flow of water and absorption of roots, nutrient removal, water temperature and food, and wildlife habitat and aesthetic values. The kit contains components for a complete demonstration of how riparian buffers affect water quality, quantity, and flow. Each kit contains 40 trees, shrubs, clay, assorted roots, flashlight, and User’s Guide.