Lab-Aids® Normal Mitosis Kit
Supplier: Lab-Aids
9
470112-508KT 297.05 USD
470112-508
Lab-Aids® Normal Mitosis Kit
Educational Materials Biology Educational Materials Cell Biology Learning Activities
Make your own mitosis microscope slides.
  • Identify the phases of plant mitosis
  • Observe chromosomal development
  • Learn micro-techniques
  • Stain and mount slides
  • Time required: one 50 minutes class period
Students prepare their own plant cells by treating scientifically grown onion root tips cells with a mild acid, then staining the cells. A stained section of cells is then squashed onto a slide and smeared into a single layer for microscopic study. All of the activities in this kit can be completed in a standard lab period.
Delivery information: See datasheet for contents information.