Lab-Aids® Bacteria Study Kit
Supplier: Lab-Aids
160
470112-506KT 167.85 USD
470112-506
Lab-Aids® Bacteria Study Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Conduct bacterial experiments in a self-paced activity.
  • Prepare and stain bacterial smears
  • Observe and identify bacteria
  • Time required: one 50 minutes class period
Students conduct the classic Leeuwenhoek bacteria experiment in this individualized, self-paced kit. Paralleling methods performed over 300 years ago, students observe and identify bacteria using a microscope. Students ultimately observe and identify bacteria through a microscope (required, but not included). Bacteria is cultured by peppercorn, hay and bean infusions prepared by the student and then mounted and gram stained on slides.
Included: student worksheets and an instructors manual
Materials needed but not supplied: microscopes
Delivery information: See datasheet for contents information.