Student Grade Economy Glass Cover Slips
Supplier: VWR International
BI0094APK100
470112-504PK 6.9 USD
470112-504
Student Grade Economy Glass Cover Slips
Cover Glasses
Economical glass cover slips for the classroom
  • Stock up on slide making supplies
Low-cost glass cover slips for student use. Available in two sizes but only one thickness, No. 1. (0.13 – 0.17 mm thick). Packages of 100.