Natural Crystal Collection
Supplier: SCOTT RESOURCES 2, LLC SE
2305
470112-330KT 57.2 USD
470112-330
Natural Crystal Collection
Specimens Mineral Specimens Mineral Collections
Identify all six crystal systems (cubic, tetragonal, orthorhombic, monoclinic, triclinic and hexagonal) with the 15 representative specimens of natural crystals included in this collection. In compartmented box with key sheet listing the locality and crystal form of each specimen.