Lab-Aids® Simulated Blood Typing Kit
Supplier: Lab-Aids
1-S
470111-910KT 113.75 USD
470111-910
Lab-Aids® Simulated Blood Typing Kit
Educational Materials Biology Educational Materials Blood Typing Learning Activities
Simulated blood typing for safety.
  • Perform blood typing without human blood
  • Observe agglutination and typing Of A, B, Ab and O blood groups
  • Time required: one 50 minutes class period
Simulated blood typing kit lets students practice the typing of A, B, AB and O blood groups while avoiding the risks associated with real blood. The clumping of simulated blood cells by corresponding simulated antibodies is done through chemical precipitation. Students observe the antigen-antibody relations of artificial bloods and sera. No human animal blood or blood products are used because the reactions are based on chemical solubility.
Included:
LAB-AIDS Slide Guides®
mixing spatulas, blue
mixing spatulas, yellow
bottle simulated Anti-A serum
bottle simulated Anti-B serum
bottle simulated Blood Type A
bottle simulated Blood Type B
bottle simulated Blood Type AB
bottle simulated Blood Type O
Student Worksheet and Guides
Teacher's Guide
SDS (Safety Data Sheet) packet
Delivery information: See datasheet for contents information.