EDVOTEK PCR EdvoBeads™
Supplier: Edvotek
625
470111-884PK 61.3 USD
470111-884
EDVOTEK PCR EdvoBeads™
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Simply add your primers and template DNA to a single PCR EdvoBead™ and mix!
  • Each vial contains enough beads for 25 PCR reactions
  • Stores in ambient temperature
PCR EdvoBead™ provide the reagents for 25 PCR reactions in a convenient ambient-temperature-stable bead. PCR Beads have been optimized for PCR reactions and contain buffer, nucleotides and Taq DNA Polymerase. The only reagents that must be added to the reaction are template DNA and specific primers.