UltraSpec-Agarose, melt and pour
Supplier: Edvotek
601
470111-866EA 39 USD
470111-866
UltraSpec-Agarose, melt and pour
Electrophoresis Gels Agarose and Acrylamide
Simply melt in microwave, water bath or autoclave; cool and pour into gel bed (400 ml—six standard gels).