Kemtec® Urinalysis
Supplier: Kemtec
8-306
470111-836EA 293.05 USD
470111-836
Kemtec® Urinalysis
Educational Materials Biology Educational Materials Physiology Learning Activities
Simulated urine testing.
  • No real or human products
  • Learn basic testing procedure
Urinalysis introduces students to to the world of biomedical technology. Using simulated samples, students evaluate the general characteristics of urine, observe for sediments, and test for glucose, albumin, pH, specific gravity, and lead.
Delivery information: See datasheet for contents information.