Lab-Aids® Genetics Concepts Kit
Supplier: Lab-Aids
70
470111-672KT 161.2 USD
470111-672
Lab-Aids® Genetics Concepts Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
Simulate multiple generations of Mendellian inheretance.
  • Rediscover Mendel's law of genetics
  • Simulate gametogenesis and fertilization
  • Reusable
  • Time required: three to five 50 minutes class periods
Duplicate Mendel’s years of work and research in five lab periods, is it possible? Yes! Through a series of simulated experiments, students experience the same problems encountered by the 'Father of Genetics'. Students 'rediscover' his Law of Genetics using specially designed GAM-ETO-DISCS® and TETRADICE® to simulate Gametogenesis and fertilization. These unique LAB-AIDS® items produce random genotype combinations that are statistically consistent. As consistent as if done with living organisms. This lab activity was developed in a game format to achieve a high level of interest and involvement by the students. All material in this activity is for 50 students is reusable.
Delivery information: See datasheet for contents information.