Lab-Aids® Human Senses Experiment Kit
Supplier: Lab-Aids
8
470111-668KT 194.35 USD
470111-668
Lab-Aids® Human Senses Experiment Kit
Educational Materials Biology Educational Materials Physiology Learning Activities
Investigate all five senses in one kit.
  • Determine olfactory fatigue and recovery times
  • 'Map' a section of skin
  • Determine the two-point threshold
  • Time required: one 50 minute class period
This kit provokes a personal interest as students identify response areas on their bodies. They become aware of their sensory perceptions and the physiological characteristics of the body receptors.
Included:
Teacher's Guide with SDS
Student Worksheets and Guides
Cotton-tipped swabs
Blind spot determiners
Metal probes
Taste solution dishes
Styrofoam ball pin holders
Bitter Solution
Salty solution
Sour solution
Sweet solution
Perfume
Oil of Cloves
Oil of peppermint
Straight pins
Bristles
Inhalers
Delivery information: See Datasheet for contents information.