Snellen's Eye Test Charts
Supplier: Able Printing Service
3539
470111-318PK 20.3 USD
470111-318
Snellen's Eye Test Charts
Charts
This sturdy chart tests for astigmatism, myopia, or hyperopia.
  • Printed in black on heavy white card stock
The chart has an astigmatism test pattern on top and a series of letters in graduated sizes with distance scales for determining myopia or hyperopia. Packaged in a set of six.