Lab-Aids® Food Nutrient Analysis Kit
Supplier: Lab-Aids
6A
470111-220EA 177.65 USD
470111-220
Lab-Aids® Food Nutrient Analysis Kit
Educational Materials Biology Educational Materials Biochemistry Learning Activities
This kit identifies five classes of nutrients.
  • Activities: Testing common foods and discussing conclusions
  • Students quantitatively determine nutrients in foods
  • Estimated lab time: one 50-minutes class period
  • Team-centered activity
In this kit, students perform and standardize tests with pure samples of nutrients. Working in teams, students identify and test for five classes of nutrients: sugar, starch, lipid, protein, and Vitamin C.
Once students complete their tests and make observations, they discuss their conclusions on the results, controls, and samples that showed no reaction.
Ordering information: This kit includes a teacher's guide with SDS, student worksheets and guides, glucose, vitamin C, protein, starch suspension, vegetable oil, fat-testing solution, protein testing solution a, protein testing solution b, sugar-testing solution, vitamin c-testing solution, starch-testing solution, and test tubes. A hot-water bath, a large sheet of cardboard or dry-erase board, and a white screen are needed but not included.
Delivery information: See datasheet for contents information.