Small Loudspeaker Set
Supplier: SARGEANTS WHOLESALE BIOLOGICAL
WL2135J
470111-130KT 108.09 USD
470111-130
Small Loudspeaker Set
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Best Seller!
Set of two 75mm diameter speakers for use in acoustics experiments such as interference. The stand is designed to avoid concealing the speaker and obstructing the sound propagation from the cone.
Set of two.