Thermometer Only
Supplier: Peco Sales
89291
470110-970EA 57 USD
470110-970
Thermometer Only
Thermometers Analog Thermometers
Permacolor-filled thermometer features projection at top to fit positioning hole.
For replacement of either wet or dry bulb thermometer in Sling Psychrometer. Permacolor-filled thermometer features projection at top to fit positioning hole.