Electric Gyrospinner
Supplier: COOK IRON STORE
100-N/7
470110-846EA 290.7 USD
470110-846
Electric Gyrospinner
Educational Materials Physics Educational Materials Mechanics Learning Activities
Spins Your Gyroscope to High Speeds Quickly and Reliably.
  • This gyrospinner turns at 5,000 and 10,000 rpm and can be used to spin all your demonstration gyroscopes
The electric gyrospinner consists of a special universal motor with a mandrel and driving disk and a molded rubber O-ring mounted on the shaft. Note: Must remove abrasive cover from the spindle before use.