Metric and English Plastic Ruler
Supplier: Acme United
M-109
470110-710CS 637.2 USD
470110-710
Metric and English Plastic Ruler
Rulers
Dual calibrated scale in center for direct metric-English comparison.
Equivalents table on back.Measures up to: 30 cm, 12", Graduations: cm, mm, inch, 1/16-inch