Sabouraud Dextrose Agar
Supplier: LABORATORY MEDIA CORP SE
LD150A
470109-176PK 81.95 USD
470109-176
Sabouraud Dextrose Agar
Chemicals for Science Education
For the cultivation and identification of fungi; Includes label instructions for preparation; must refrigerate upon receipt.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.