Borealene II
Supplier: ALDON CORP SE
BB0174-3.8
470108-366EA
37.85
USD
470108-366
Borealene II
Chemicals for Science Education
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Density (g/mL): <1.0
Boiling Point (°C): >100
Synonyms: Formula II Dilute (Ready to Use)
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...