Tuffy Carts
Supplier: H. WILSON COMPANY
WT34E WT26CE WT34CE WT42CE WT34E WT26CE WT34CE WT42CE
470107-834EA 25.25 USD
470107-834 470107-836 470107-838 470107-840 470113-994 470113-998 470114-000 470114-002
Tuffy Carts
Carts
Constructed from an engineered thermoplastic resin, these durable laboratory carts will not chip, warp, crack, rust or peel.
All shelves feature a raised textured surface to prevent materials from slipping while cart is in motion. Included with every color Tuffy cart is a three outlet UL/CSA electrical assembly with a 15 ft. cord, cord management wrap and three cable management clips. Electrical attachment is recessed so it doesn't add to the width of the cart. The cart cabinets are constructed of 20 gauge steel with key locks.
Ordering information: Available in red, blue or black. Please specify color choice when ordering.
Delivery information: All sizes except for 77004-81 come with cabinet.