Coliscan® EasyGel® Water Monitoring Kit
Supplier: MICROLOGY INC
CWM10
470107-534KT 133 USD
470107-534
Coliscan® EasyGel® Water Monitoring Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Make Sure the Water is Free of E. coli
  • Collect local water samples
  • Linked to a real world scenario
  • Includes an instruction guide
Coliscan Water Monitoring Kit features Easygel, an agar replacement. Each kit has materials for 10 tests. The kit includes Coliscan Easygel bottles, Easygel petri dishes, 30mL collection bottles, 1mL Droppers, instruction guide, and Coliscan Interpretation Guide.