Mini Nutating Rocker, 230 V
Supplier: Ward's
H3D-1020-E
470107-128EA 594.6 USD
470107-128
Mini Nutating Rocker, 230 V
Shakers and Mixers Rotating/Rocking Shakers
This mini mixer takes up minimal bench space, while providing maximum mixing efficiency.
  • 230 V model
  • Fixed speed
  • Small platform
  • Flat cord stays out of your way
  • Two year warranty
Capable of supporting loads of up to four pounds, this fixed speed orbital mixer is small but powerful. With 3D nutating movement, it will agitate your samples gently, but thoroughly.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (470107-056) is also available, please see separate product page.