Plastic Four-Sided Meter Stick
Supplier: SI Manufacturing
45780-A
470106-452EA 21 USD
470106-452
Plastic Four-Sided Meter Stick
Rulers
Four meter sticks in one and made of flexible plastic.
The sides have 1 m, 10 dm, 100 cm, and 1000 mm graduations.