Soil Thermometer
Supplier: Taylor Precision Products
5976N
470106-366EA 11.2 USD
470106-366
Soil Thermometer
Thermometers Analog Thermometers
Determine Soil Temperature Quickly.
  • Large visible display of optimal growing conditions
  • Durable
  • Aligned with GLOBE protocols
Use this stainless steel thermometer to test soil temperatures for planting. The dial, which is 6 cm in diameter, records temperatures from 0 to 80 °C and 20 to 180 °F. The 12.5 cm stem is long enough for most planting depths.