Frigimat® Cub Dry Ice Maker
Supplier: Bel-Art
F38874-0000_SE
470106-034EA 785.5 USD
470106-034
Frigimat® Cub Dry Ice Maker
Ice Makers
Create dry ice on demand. Attach this compact unity to a liquid CO2 cylinder and in a few minutes you'll have a ready-to-use 250 - 350 g block of dry ice.
  • Dry ice production can be viewed through a translucent polycarbonate body that opens without clamps or screws
  • System operates multiple pressure relief safeguards
Size: 170x190x150 mm.
Weight: 4.8 lbs.