Glass Thermometers
Supplier: Sper Scientific
738730
470105-948PK 131.5 USD
470105-948
Glass Thermometers
Thermometers Analog Thermometers
All filled with safe red alcohol, white-backed, total immersion, 6" (153 mm) long, in blue plastic holder.
  • Pack of 12
  • Pocket thermometer