Properties of Ionic, Molecular and Metallic Solids Lab Activity
Supplier: Avantor
470105-836EA 199.79 USD
470105-836
Properties of Ionic, Molecular and Metallic Solids Lab Activity
Educational Classroom Kits and Activities
Learn How to Make a Conductivity Indicator
  • Hands-on class activity
  • Great for grades 8-11
Teacher Angela Riordan created this interesting hands-on lab activity that demonstrates how properties of substances can be used for identification and classification. Students test samples of different solids and then determine whether they are ionic, molecular or metallic solids from their physical properties such as hardness, melting point, solubility in polar and non-polar solvents, and electrical conductivity.
Ordering information: A conductivity indicator is required, but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout