Burette Clamps
Supplier: United Scientific Supplies
CLBUR2N
470105-692EA 26.75 USD
470105-692
Burette Clamps
Clamps General Purpose Clamps
Easily hold two burettes at once.
  • PVC coated jaws
  • Accepts burettes up to 35 mm O.D.
Aluminum double clamp is designed to securely hold burettes up to 35 mm in diameter. Spring jaws are covered with non-slip PVC sleeves. Compatible with rods up to 13 mm in diameter.