Laboratory Chow
Supplier: INDUSTRIAL SUPPORT INC.
21-1926
470105-576PK 73.75 USD
470105-576
Laboratory Chow
Animal Supplies Animal Feed
Use this complete diet for mice, gerbils, rats and hamsters.
The average monthly feed consumption for each animal is as follows: mouse - 150 g; gerbil - 200 g; rat - 450 g; and hamster - 300 g. Includes 5kg.