Minit® DNA Model
Supplier: Cochranes of Oxford
0095
470105-550EA 239 USD
470105-550
Minit® DNA Model
Models Botany Models
An impressive molecular biology teaching resource and display model.
  • Aids in understanding of DNA structure
  • Assembly engages students
  • Connectors are differentiated for hydrogen bonding
Building the DNA model engages and educates students. Groups can work together to make up the subassemblies – 15 Thymine-Adenine and Cytosine-Guanine base pairs, sugar rings and phosphate groups – which are joined together to make one and a half turns of the double helix and to show the major and minor grooves of the protein lysozyme.
Each atom is shown, color-coded,More Product Information