Tubby® Storage Container
Supplier: Heathrow Scientific
HS23453
470105-518CS 337.7 USD
470105-518
Tubby® Storage Container
Boxes Storage Boxes
Tubby® stackable storage container is designed to help organize a laboratory.
  • The lid snaps in place to secure the contents and for safe transport
  • Constructed of high-impact polypropylene
The removable dividers can create as big or as small a compartment as needed. Perfect for keeping tubes, tips, pipettes, gloves and other small items organized and out of the way.