Glow Chemistry Kit II: It’s Blue!
Supplier: Avantor
470105-492EA 118.09 USD
470105-492
Glow Chemistry Kit II: It’s Blue!
Educational Classroom Kits and Activities
Chemistry teacher, Stephen Freedman, uses this demonstation at Halloween.
  • Grades 8 to 12
Two solutions, A and B, are created with ammonium bicarbonate, hydrogen peroxide, sodium bicarbonate, sodium carbonate luminol and copper II sulfate. Mix the solutions together and a dramatic blue glow appears, which is very eerie, especially in a darkened room.
Ordering information: Needed but not included is distilled water. Includes 10 demonstrations and instructions.
Caution: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.