Hanna Instruments Iron (Fe²⁺and Fe³⁺) Chemical Test Kit
Supplier: Avantor
470105-390KT 26 USD
470105-390
Hanna Instruments Iron (Fe²⁺and Fe³⁺) Chemical Test Kit
Environmental Test Kits
This chemical test kit that uses the phenanthroline method to measure total iron in the medium range. This chemical test kit is supplied complete with all of the reagents and equipment necessary to perform approximately 50 tests.
  • Pre-made reagents for ease of use
  • Color comparison cube for simple determination
  • Complete setup: All required materials are included with the test kit, such as the sample beaker, color comparison cube, and reagent packets
  • High resolution: Readings from 0 to 5 mg/L are determined to 1 mg/L resolution
Iron is naturally present in water in low concentrations, but it reaches high concentrations in wastewater effluents. The iron concentration in water needs to be monitored because it becomes harmful above certain levels. In domestic water, for instance, iron can stain laundry, damage kitchenware, favor the growth of certain bacteria, and unpleasantly alter the taste of water. Iron is also an indicator of ongoing corrosion in water cooling and heating systems. Moreover, iron is normally monitored in mining wastewater to avoid contamination.
Ordering information: Replacement reagents need to be purchased seperatly.