Ceramic Ring Magnets
Supplier: Master Magnetics
CR162
470105-290PK 17.6 USD
470105-290
Ceramic Ring Magnets
Magnets
These ceramic ring magnets have an inside diameter of 35 mm and an outside diameter of 45 mm.
They are approximately 6.4 mm thick and are magnetized parallel with the thickness.
Packaging: Package of 16.