Permanent Ceramic Disc Magnet
Supplier: Master Magnetics
CD022800
470105-152EA 5.15 USD
470105-152
Permanent Ceramic Disc Magnet
Magnets
A permanent ceramic disc magnet with the North and South poles on opposite ends of the diameter.
58 mm diameter and 6.4 mm thick.