Permanent Ceramic Ring Magnet
Supplier: Master Magnetics
CR238128/6
470105-150EA 0.63 USD
470105-150
Permanent Ceramic Ring Magnet
Magnets
A permanent ceramic ring magnet with multiple poles on the faces.
The center hole is 25 mm. Shown with our Permanent ceramic disc magnet. Size: 60 mm diameter and 6 mm thick.